missing translation for 'onlineSavingsMsg'
Learn More

Ansell MICROFLEX™ UltraSense™ EC USE-880 Nitrile Gloves
GREENER_CHOICE

Catalog No. 19167295A
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19167295A Small
19167295F X-Small
19167295B Medium
19167295D X-Large
19167295C Large
19167295E 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19167295A Supplier Ansell Supplier No. USE880S
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Extended cuff offers extra protection over the wrist and forearm for use in high-risk environments.

High-quality nitrile offers reliable barrier protection against many hazards and infectious substances.

  • Patented ultra-soft nitrile provides the fit and feel of latex
  • Textured fingers ensure a secure grip
  • Extended, beaded cuff offers extra protection over wrist and forearm
  • Nonsterile, ambidextrous, single-use
  • Length: 11.4 in. (29 cm)
  • Fingertip thickness: 4.3 mil

Specifications

Certifications/Compliance ASTM D 6319
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Chemotherapeutic Rated No
Chlorinated Yes
Applications Healthcare, Laboratory
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 5.1 mil
Finger Thickness (Metric) 0.13 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 3.9 mil
Palm Thickness (Metric) 0.10 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Small
Sterility Non-sterile
Thickness (English) 5.1 mil
Thickness (Metric) 0.13 mm
FDA 510 (k) Yes
Glove Cuff Style Beaded
Model USE-880-S
Quantity 1000/Cs.
Series UltraSense™ EC USE-880
Tensile Strength 33 MPa
Length (English) 11.4 in.
Length (Metric) 29 cm
Show More Show Less