missing translation for 'onlineSavingsMsg'
Learn More

Ansell MICROFLEX™ MIDKNIGHT™ XTRA 93-862 Nitrile Gloves with Extended Cuff
GREENER_CHOICE

Catalog No. 19500805
Change view
Click to view available options
Size:
Large
Medium
Small
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19500805 Medium
19500804 Small
19500806 Large
19500807 X-Large
19500808 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19500805 Supplier Ansell Supplier No. 93862080
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Disposable nitrile gloves with extended cuff provide durable protection in the event of chemical splashes. NFPA and medical exam grade certified for use by healthcare professionals.

  • Tested against both fentanyl and gastric acid (vomit) to simulate authentic overdose situations
  • Dark, black color makes it easy to see when hands have come into contact with potentially hazardous powders
  • Made with a soft, flexible nitrile formulation
  • Fully textured for a reliable, consistent grip
  • For use with:
    • Administering drugs
    • Emergency services
    • Equipment repair and maintenance
    • Extra protection over wrist and forearm
    • Inspection, selecting, checking parts
    • Laboratory analysis and testing
    • Paint and spray shops
    • Patient examinations
    • Raw material sample collection
    • Tire rotation and change

Specifications

Certifications/Compliance NFPA 1999:2018, ASTM D6319, FDA21 CFR 177-2600
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, EN 455 1-4
Accelerator Free No
Applications Healthcare, Laboratory
AQL 0.65
Color Black
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 6.3 mil
Finger Thickness (Metric) 0.16 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4.7 mil
Palm Thickness (Metric) 0.12 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Medium
Sterility Non-sterile
Thickness (English) 6.3 mil
Thickness (Metric) 0.16 mm
Model 93862080
Series MICROFLEX™ MidKnight™ XTRA 93-862
Length (English) 12 in.
Length (Metric) 30 cm
Show More Show Less