missing translation for 'onlineSavingsMsg'
Learn More

Ansell MICROFLEX™ MEGA TEXTURE 93-283 Blue Gloves

Blue nitrile disposable gloves are strong and durable to enable workers to confidently grip oily, fatty or wet foods and objects.

Supplier:  Ansell 93283080

 View more versions of this product

Catalog No. 19-500-870



Only null left
Explore available promotions

Description

Description

  • Mega Texture surface for secure grip when handling slippery objects
  • Generous fit works well with thermal under gloves
  • Cut-protective design resists tearing
  • Compliant with US Food Contact and EU Food Migration Regulations
Specifications

Specifications

ASTM D6319, FDA 21 CFR 177-2600, ISO 9001, ISO 13485
No
Industrial
Blue
Fully Textured
Nitrile
Nitrile Gloves
Non-sterile
0.22 mm
MICROFLEX™ Mega Texture 93-283
11.8 in.
EN 374-1:2016, EN 420:2003 + A1:2009, EN 374-5:2016
No
1.5
Single-use
Yes
Powder Free
Medium
8.7 mil
No
28 MPa
300 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions