missing translation for 'onlineSavingsMsg'
Learn More

Ansell MICROFLEX™ Lifestar EC™ LSE-104 High Risk Nitrile Gloves
GREENER_CHOICE

Catalog No. 19010320
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19010320 3X-Large
19010315 Small
19010316 Medium
19010317 Large
19010318 X-Large
19010319 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19010320 Supplier Ansell Supplier No. LSE1043XL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Disposable EMS gloves provide protection against bloodborne pathogens, chemical splashes and other hazardous substances. Dual layer, dual color, high risk exam gloves are specially designed for use while caring for patients, administering drugs, and performing other vital tasks.

  • Designed specifically for EMS and healthcare professionals
  • Durable for safe critical patient care
  • Dual layer, dual color design for two layers of protection
  • Bright blue interior makes breaches more detectable
  • White exterior makes fluid and blood contact more visible
  • Tested against both fentanyl and gastric acid to simulate hazardous, real world overdose situations
  • Advanced barrier protection (AQL 0.65)
  • Non-stick and non-foaming properties for reduced interference when working

Specifications

Certifications/Compliance ASTM D6319, FDA21 CFR 177-2600, ISO 13485, ISO 9001, NFPA 1999:2013
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Chemotherapeutic Rated No
Chlorinated Yes
Accelerator Free Yes
Applications Healthcare
AQL 0.65
Color White, Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 8.3 mil
Finger Thickness (Metric) 0.21 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5.5 mil
Palm Thickness (Metric) 0.14 mm
Powdered Powder Free
Size 3X-Large
Sterility Non-sterile
Thickness (English) 5.5 mil
Thickness (Metric) 0.14 mm
FDA 510 (k) No
Glove Cuff Style Beaded
Model LSE-104-3XL
Quantity 900/Cs.
Series MICROFLEX™ Lifestar EC™ LSE-104
Tensile Strength 25 MPa
Length (English) 11 in.
Length (Metric) 27.9 cm
Show More Show Less