missing translation for 'onlineSavingsMsg'
Learn More

Microcrystalline Cellulose, NF, Spectrum™ Chemical

Catalog No. 18604591
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604591 100 g Poly Bottle
18604592 500 g Poly Bottle
18604593 2.5 kg Poly Pail
18604595 12 kg Fiber Drum
18604594 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604591 Supplier Spectrum Chemical Mfg Cor Supplier No. C1679100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

C1679, 9004-34-6

Spectrum™ Chemical Microcrystalline Cellulose, NF is an excipient in pharmaceutical formulations acting as a binder in tablet formation. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-34-6
Molecular Formula C14H26O11
Molecular Weight (g/mol) 370.35
InChI Key PTHCMJGKKRQCBF-UHFFFAOYNA-N
IUPAC Name 2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol
SMILES COC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O

Specifications

CAS Max % 100%
Quantity 100 g
Grade NF
Ignition Residue 0.1%
Loss on Drying 7%
Packaging Poly Bottle