Learn More
Kayaku Advanced Materials MICROCHEM Remover PG 4L DFS Item
Supplier:  Microchem Corp G0502004000L1PE
For single bottles only. To order 4/case, please see item NC1258145. MicroChem Remover: Clean Efficient Removal - Residue-free removal with less agitation. AQUEOUS ALKALINE, METAL ION FREE. Reduced device contamination. Rinses completely in water. No strong objectionable odor; Simple replenishment; Cost effective two bath system
Catalog No. NC9893682
Only null left
Explore available promotions