missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Microchem I Printer Paper for SafTest™ Test Kits

Catalog No. MP07MC1030
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP07MC1030 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP07MC1030 Supplier MP Biomedicals™ Supplier No. 07MC1030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Microchem I Printer Paper for Saftest™ analyzer.

  • Accessory for SafTest™ vacuum pump
  • Replacement labware
  • Size: 6.35 x 2434.8 cm (2.5 x 960 in.)
  • Format: Roll

Specifications

Quantity 1 Ea.
Storage Requirements Room temperature