missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 Ts Plus Coveralls
Breathable, microporous laminate suit protects against liquid and particulate biological hazards including radioactive particulates. Previously known as MICROCHEM™ by AlphaTec 2000 STANDARD.
Supplier: Ansell WH20T9211108
Description
- Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Anti-static, tested according to EN 1149-5 and AATTC 76
- Ultra Low Linting, reduced risk of contamination in critical areas
- Silicone free, critical in spray painting applications
- Elasticated hood, wrists, and ankles help to minimize the risk of linting and cross contamination
- Free of natural rubber latex
Specifications
| General Purpose Coveralls | |
| Unisex | |
| White | |
| Elastic | |
| Thumb Loop | |
| 3-panel, Respirator Fit | |
| Serged, Taped | |
| Liquid, Particle, Bloodborne Pathogens | |
| CE CATEGORY III (Type 4-B, Type 5-B, Type 6-B, EN 1073-2, EN 14126, DIN 32781, EN 1149-5) | |
| WH20-T-92-111 | |
| Life Sciences, Automotive, Metal Fabrication, Food Processing, Utilities, Oil and Gas, Machinery, Chemical |
| 4X-Large | |
| Microporous Polyethylene Laminate (Non-woven) | |
| Limited-use | |
| Elastic | |
| Hooded | |
| Front Zipper with Storm Flap | |
| Non-sterile | |
| Hooded | |
| Yes | |
| None | |
| 25/Cs. |