missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 2000 Shoe Covers - Model 417

Catalog No. p-7218237
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18167007 Large
18167008 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18167007 Supplier Ansell Supplier No. WH20B9241700
Only null left

Breathable microporous laminate technology provides protection from low hazard liquid spray and fine particulates.

  • Proven protective barrier to low concentration liquid chemicals, liquid & particulate biological hazards
  • Comfortable to wear featuring moisture vapor permeable fabric to help reduce the risk of heat stress
  • Optimized fit, improving wearer comfort and safety
  • Low Linting

Specifications

Size Large
Material Microporous Polyethylene
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top
Sole Type ESD PVC
Model 417
Seam Type Bound
Style AlphaTec™ 2000
For Use With (Application) Life Sciences, Metal Fabrication, Food Processing
Certifications/Compliance (Europe) EN ISO 13688:2013
Product Type Shoe Covers
Show More Show Less