Learn More
Brands MICROCELL CARY 50 40UL 2-1/UN
Catalog No. 500111594
Encompass
Agilent LS Microcell Cary 50 40ul, 2mm x 10mm, 1/p V UV Cuvettes-58 58
Catalog No. 500111594 Supplier Brands Supplier No. 6610019500
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Agilent LS Microcell Cary 50 40ul, 2mm x 10mm, 1/p V UV Cuvettes-58 58