missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Full-Skirted Plates with Barcode, red

Catalog No. A31729
Encompass
Change view
Click to view available options
Quantity:
50 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31729 50 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A31729 Supplier Applied Biosystems™ Supplier No. A31729
Only null left

The Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Full-Skirted Plate is optimally designed for use with the Applied Biosystems™ Automated Thermal Cycler (ATC).

These full-skirted reaction plates are manufactured using a durable polycarbonate frame, making them ideal for easy handling with robotic arms and other automated system features. The thin-wall construction of the plate allows for efficient heat transfer and optimal fit in thermal cycler blocks. The black alpha- numeric labeling provides easy-to-read well identification during sample preparation. The MicroAmp EnduraPlate Optical 96-Well Full-Skirted Plate is the only 96-well plate recommended for use with the Automated Thermal Cycler (ATC) in the automated PCR workflow process.

Key features include:
• Designed exclusively for use with Automated Thermal Cycler (ATC) (Cat No. A31486)
• Full-skirted polycarbonate frame for easy robotic handling and plate stacking/loading
• 3-way barcodes on alpha, opposite alpha, and opposite numeric sides to improve plate reading efficiency
• Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and automated work processes
• Certified free of DNA, RNase, and PCR inhibitors
• Available in multiple colors for easy organization of laboratory assays
• Excellent mechanical stability and flatness for your complete automated workflow needs

Specifications

Barcode Includes Barcode
Certifications/Compliance Certified DNA/RNase Free
Content And Storage Store at room temperature.
Packaging 50 Plates
Product Line MicroAmp
Color Optical, Red
Product Type Microplate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style Skirted
Material Polycarbonate, Polypropylene
Volume (Metric) Well 0.2 mL
No. of Wells 96
Volume (Metric) Working 0.1 mL
Total Volume (Metric) Well 0.2 mL
Quantity 50 plates
Shipping Condition Room Temperature
Show More Show Less