missing translation for 'onlineSavingsMsg'
Learn More

Ansell MICRO-TOUCH™ NITRAFREE™ Blue

Catalog No. 17000590
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17000590 Large
17000588 Small
17000589 Medium
17000587 X-Small
17000591 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17000590 Supplier Ansell Supplier No. 313028090

MICRO-TOUCH™ NITRAFREE™ - Nitrile, examination glove with reliable protection

  • Manufactured without sulfur-based chemical accelerators, helping to protect users from type I latex allergies and type IV chemical allergies.
  • Thin and light weight formulation provides enhanced tactile sensitivity and comfort for longer wear time.
  • Unique berry blue color provides contrast, while masking blood and other stains.
  • Tested for use with chemotherapy drugs.

Specifications

Chemotherapeutic Rated Yes
Chlorinated No
Accelerator Free Yes
Applications Healthcare
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 3.9 mil
Finger Thickness (Metric) 0.10 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 2.8 mil
Palm Thickness (Metric) 0.07 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Large
Sterility Non-sterile
FDA 510 (k) Yes
Glove Cuff Style Beaded
Quantity 2000/Cs.
Series MICRO-TOUCH NITRAFREE™ Blue
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less