missing translation for 'onlineSavingsMsg'
Learn More

Basix™ Micro Tips, Universal Fit
GREENER_CHOICE

Catalog No. 13611103
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
Refill
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Format
13611103 Non-sterile Bulk
13611107 Sterile Rack
13611106 Non-sterile Rack
13611108 Non-sterile Refill
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13611103
Only null left
Edge

The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.

The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.

Basix offers reliability and quality for all varieties of experiments, from everday liquid transfer to PCR and Electrophoresis, Basix offers a tip to meet your needs.

Choose from non-Sterile or Sterile with packaging options including bulk, racked, reloads, stack racks.

  • All racked sterile tips are shrink wrapped and certified free of RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors and endotoxin and DNA
  • All nonsterile racked and bulk nonfiltered tips are certified free of RNase, DNase, and DNA

Specifications

Volume (Metric) 10 μL
Sterility Non-sterile
Color Natural
Length (English) 1.25 in.
Length (Metric) 3.17cm
Clean Claims DNase, RNase, and DNA Free
Format Bulk
Tip Style Micro Point, Universal
For Use With (Equipment) Universal Fit