missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Micro Tip Box, Empty

Catalog No. S89608
Change view
Click to view available options
Size:
2 to 200 μL
200 to 1000 μL
Length (Metric):
126 mm
143 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Length (Metric)
S89608 200 to 1000 μL 143 mm
S89607 2 to 200 μL 126 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S89608 Supplier United Scientific™ Supplier No. P20103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

United Scientific™ Micro Tip Boxes come in a pack of six empty micro tip boxes and hold up to 96 tips.

Pack of six empty micro tip boxes; each box holds up to 96 tips 2 to 200 μL or 200 to 1000 μL capacity.

  • Offered at 2 to 200 μL or 200 to 1000 μL tip capacity
  • Holes are uniformly spaced for use with a multi-channel micropipette
  • Transparent top covers included
  • Autoclavable

Specifications

Autoclavable Autoclavable
Color Blue/Clear
Holds Micropipette Tips (1000μL)
Material Polypropylene
Positions 96
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Pipette Tips
Size 200 to 1000 μL
Length (Metric) 143 mm
Includes Empty Micro Tip Box with Cover
Product Type Pipette Tip Rack
Array 12 x 8
Show More Show Less