Electron Microscopy Sciences Micro-Test/Staining Dish
Catalog No. 50931200
This staining dish is made from clear glass and has 10 cells in 2 rows of 5 each. Each cell is 2 mm deep and holds 0.15 ml of solution. Very useful in specimen preparation, EM staining, and Boerner-Jones-Lukens microfluocculation test.
Catalog No.
50931200
Supplier
Electron Microscopy SciencesSupplier No.
71564
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This staining dish is made from clear glass and has 10 cells in 2 rows of 5 each. Each cell is 2 mm deep and holds 0.15 ml of solution. Very useful in specimen preparation, EM staining, and Boerner-Jones-Lukens microfluocculation test.