missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micro Particles, Magnetic, Streptavidin coated, 1 μm Particle Size, Std dev <0.1 μm, MilliporeSigma™ Supelco™
Specifications
Specifications
| Concentration | 1% (solids) |
| Content And Storage | 2°C to 8°C |
| Quantity | 5 mL |
| Synonym | Polystyrene beads, magnetic, streptavidin coated |