missing translation for 'onlineSavingsMsg'
Learn More

Micro Particles Based on Polystyrene, Size: 5 m, MilliporeSigma™ Supelco™

$634.68 - $912.53

Specifications

Concentration 10% (solids)
Content And Storage 2°C to 8°C
Synonym Latex beads from PS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
111004010
View Documents Promotion Details
N/A 5 mL
Each for $634.68
Only null left
 
111003777
View Documents Promotion Details
N/A 10 mL
Each for $912.53
Only null left
 
Description

Description

Specifications

Specifications

10% (solids)
2°C to 8°C
Latex beads from PS
Videos
SDS
Documents

Documents