missing translation for 'onlineSavingsMsg'
Learn More

Micro Essential Lab pHIZATEST™ Nitrazine Indicator Paper

Catalog No. 14850115
Change view
Click to view available options
pH Range:
4.5 to 7.5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
14850115 4.5 to 7.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14850115 Supplier Micro Essential Lab Supplier No. 934
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phenaphthazine 4.5–7.5 nitrazine indicator paper has been trusted by hospitals and physicians worldwide for more than 50 years.

  • The economical alternative to higher priced nitrazine papers
  • Test papers give instant, easy-to-read results, with clear, bright matches at 0.5 intervals from pH 4.5 to 7.5
  • Color charts are lab-tested for precision accuracy
  • An FDA-listed medical test product, used in hospitals and doctor’s offices around the world
  • Made in the U.S.A.
  • Each kit consists of a 15 ft. x 1/4 in. roll of premium test paper in a handy dispenser with matching color chart

Specifications

Color Indicator Nitrazine
pH Range 4.5 to 7.5
Graduations 0.5
Quantity 100 Tests
Type Nitrazine Indicator Paper