Learn More
Micro Essential Lab Mikro Hydrion™ pH Test Paper
Supplier: Micro Essential Lab M1000
Specifications
| 0 to 14 | |
| Two papers in dispenser pH range 0-11 (graduations 1) and 12-14 (graduations 0.5) | |
| Each | |
| Hydrion™ Micro Dispenser |