missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micro Essential Lab Hydrion™ Single Roll Dispenser, 4.8 to 6.7 pH
Immediate, accurate response pH test strips
Supplier: Micro Essential Lab #333
Includes: Matching color chart
Description
For classroom, laboratory and general testing applications.
- Short pH Range: 4.8 to 6.7
- Distinct color matches at pH 4.8, 5.3, 5.9, 6.3 and 6.7
- 4.57m (15 ft.) long × 6.35mm (1/4 in.) wide roll provides 100 to 150 tests
Compliance
Calibration verified using NIST™ traceable standards
Specifications
| 4.8 to 6.7 | |
| 100 to 150 Tests | |
| 15 ft. x 1/4 in. (4.57 m x 6.35 mm) | |
| 4.57 m | |
| 6.35 mm |
| Color chart | |
| Single Roll pH Paper Dispenser | |
| 15 ft. | |
| 1/4 in. |