missing translation for 'onlineSavingsMsg'
Learn More

Micro Essential Lab Hydrion™ Single Roll Dispenser, 2 to 10 pH

Catalog No. 14853150M
Change view
Click to view available options
pH Range:
2 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
14853150M 2 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14853150M Supplier Micro Essential Lab Supplier No. MES48
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Color chart, pH strip roll, dispenser

Immediate and accurate general use pH test strips

For classroom, laboratory and general pH testing applications.

  • Wide pH Range: 2 to 10
  • Distinct color matches every 2 pH points (2, 4, 6, 8, 10)

Compliance

Calibration verified using NIST™ traceable standards

Specifications

pH Range 2 to 10
Includes Colorimetric chart
Quantity Each
Type Single Roll pH Test Strip Dispenser
Width (English) 1/4 in.
Width (Metric) 6.35 mm