Learn More
Micro Essential Lab Hydrion™ (QK-1000) DR Quaternary Test Kit and Refill
Provides a simple and reliable means to measure the concentration of quaternary sanitizers, specifically Alkyldimethylbenzyl ammonium compounds, Rocall II and Hyamine 3500. Keep refills on hand (15-420-1W) to minimize downtime.
Supplier: Micro Essential Lab QKR1000
Description
- Color matches within the Hi Range of 0-200-400-600-800-1000ppm (parts per million) and the Lo Range of 0-100-200-300-400ppm
- Test paper measures concentrations between 0-1000ppm
- Kit detects exhaustion of solutions that should be replaced as well as helping to avoid using excessive amounts of sanitizing agents
- Refills available: 15-420-1W
Chemical Identifiers
| For concentration of quaternary sanitizers (alkyldimethyl benzyl ammonium compounds, Rocall II, Hyamine 3500) |
Specifications
| For concentration of quaternary sanitizers (alkyldimethyl benzyl ammonium compounds, Rocall II, Hyamine 3500) | |
| 5 Pack | |
| Quaternary Test Kit Refill |