Learn More
Micro Essential Lab Hydrion™ (QK-1000) DR Quaternary Test Kit and Refill
Provides a simple and reliable means to measure the concentration of quaternary sanitizers, specifically Alkyldimethylbenzyl ammonium compounds, Rocall II and Hyamine 3500. Keep refills on hand (15-420-1W) to minimize downtime.
Supplier:  Micro Essential Lab QK 1000
 View more versions of this product
Catalog No. 154201T
Only null left
Description
Description
  • Color matches within the Hi Range of 0-200-400-600-800-1000ppm (parts per million) and the Lo Range of 0-100-200-300-400ppm
  • Test paper measures concentrations between 0-1000ppm
  • Kit detects exhaustion of solutions that should be replaced as well as helping to avoid using excessive amounts of sanitizing agents
  • Refills available: 15-420-1W
Specifications
Chemical Identifiers
For concentration of quaternary sanitizers (alkyldimethyl benzyl ammonium compounds, Rocall II, Hyamine 3500)
Specifications
For concentration of quaternary sanitizers (alkyldimethyl benzyl ammonium compounds, Rocall II, Hyamine 3500)
A high range color match chart with distinct matches at increments of 0 to 1000ppm and a low range color chart with clear colors at increments of 0 to 400ppm are included
Each
Quaternary Test Kit
Product Suggestions
Product Suggestions