missing translation for 'onlineSavingsMsg'
Learn More

Micro Essential Lab Hydrion™ One Drop Concentrated Indicator Solution Kit

Catalog No. 14853163
Change view
Click to view available options
pH Range:
1 to 11
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
14853163 1 to 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14853163 Supplier Micro Essential Lab Supplier No. UI100
Only null left

Includes

  • 24mL (0.8 oz.) of pH indicator solution in polyethylene dropper bottle
  • 4mL (0.14 oz.) sample tube (capped)
  • Laminated color chart
  • Polyethylene protective case

Concentrated pH indicator solution only needs one drop to test pH

  • One drop of indicator solution added to 4mL (0.14 oz.) of sample provides accurate pH reading
  • Bright, easy to view color display remains stable for delayed readings
  • pH Range: 1.0 to 11.0
  • Color chart has distinct matches at every full pH unit

For testing pH of solutions

Specifications

pH Range 1 to 11
For Use With (Application) For testing pH of solutions
Includes 24mL of pH indicator solution in polyethylene dropper bottle , 4mL sample tube (capped) and a laminated color chart, packed in a polypropylene protective case
Quantity Each
Type pH Indicator Solution