Learn More
Micro Essential Lab Hydrion™ Jumbo Single Roll pH Test Paper Dispensers
Hydrion™ Jumbo Size pH Papers are easier to read with larger color charts. The 0.5 in. width of the paper enables clearer and quicker color matching.
Supplier:  Micro Essential Lab HJ610
 View more versions of this product
Catalog No. 14853158B
Only null left
Description
Description
Hydrion™ pH Paper is a general use pH paper. A sample of its diverse pH applications include:
  • Classroom Demonstrations
  • Hospital Laboratories
  • General Laboratories
  • Industrial Testing
  • Food Service
  • Safety Testing
  • Emergency Testing
  • Agricultural Testing
Each carton contains 10 kits, each consisting of a 15-foot roll of test paper and matching color chart. Provides for approximately 1000 tests.
Directions
Simply tear off a small strip of pH paper, dip into the test solution, then instantly compare the resulting color with the matching pH color chart.
Specifications
Chemical Identifiers
Reusable dispenser, Matching color chart, 1 roll of pH paper
15.2 m x 1.3 cm (50 ft. x 0.5 in.)
15.2 m
1.3 cm
1000 Tests
50 ft.
0.5 in.
Specifications
1 to 11
1000 Tests
15.2 m x 1.3 cm (50 ft. x 0.5 in.)
15.2 m
1.3 cm
Reusable dispenser, Matching color chart, 1 roll of pH paper
Jumbo/Wide-range pH Test Paper
50 ft.
0.5 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
This product is not intended to diagnose, treat, cure or prevent any disease or condition. Please contact a medical doctor to diagnose and treat any medical condition.