Learn More
Research Products MICRO-CENTRIFUGE TUBE CUTTER DFS Item
Supplier:  Research Products Corporation RPIF21018
Safely and easily sections microcentrifuge tubes and eliminates the hazards of using razor blades or knives. Easily retrieve radioactive pellets from the tips of tubes without resuspension or the risk of splashed radioactive substances. Works well with polyethylene or polypropylene microcentrifuge tubes up to 1.5 ml; not recommended for polystyrene tubes. Chrome-plated body with replaceable steel blade. Maximum cut diameter is 9.4 mm (0.370 inches).
Catalog No. 50998995
Only null left
Explore available promotions