Learn More
Brands Rectangular sub-micro quartz cell
Catalog No. NC1799713
Encompass
Rectangular sub-micro cell, quartz, stoppered with PTFE stopper supplied, 3 x 4.5mm aperture, 10 mm pathlength, top section of the cell features two self-masking black walls and two translucent side walls, 135 L, z height 20mm, 1/pk. Compatible with Agilent Cary 50/60 UV-Vis spectrophotometers.
Catalog No. NC1799713 Supplier Brands Supplier No. 6610021100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Rectangular sub-micro cell, quartz, stoppered with PTFE stopper supplied, 3 x 4.5mm aperture, 10 mm pathlength, top section of the cell features two self-masking black walls and two translucent side walls, 135 L, z height 20mm, 1/pk. Compatible with Agilent Cary 50/60 UV-Vis spectrophotometers.