Learn More
International Products Corp Micro-90 Concentrated Alkaline Cleaning Solution, 4x4L/case
Catalog No. NC0081746
Encompass
Micro-90 is a mild, yet powerful, multipurpose, alkaline cleaning concentrate that has long been used in laboratories, industrial applications, and critical cleaning processes. Micro-90 is a unique chelating detergent that contains ionic and non-ionic ingredients, which combine to produce a variety of cleaning actions. Micro-90 lifts, disperses, emulsifies, sequesters, suspends, and decomposes soils, then rinses away leaving the surface absolutely clean. Registered with NSF as an A1 cleaner and can be validated in FDA processes. Validation methods are available upon request. Micro-90 is manufactured in a dedicated system. Typical pH: 9.5.
Catalog No. NC0081746 Supplier International Products Corp Supplier No. M905104
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Micro-90 is a mild, yet powerful, multipurpose, alkaline cleaning concentrate that has long been used in laboratories, industrial applications, and critical cleaning processes. Micro-90 is a unique chelating detergent that contains ionic and non-ionic ingredients, which combine to produce a variety of cleaning actions. Micro-90 lifts, disperses, emulsifies, sequesters, suspends, and decomposes soils, then rinses away leaving the surface absolutely clean. Registered with NSF as an A1 cleaner and can be validated in FDA processes. Validation methods are available upon request. Micro-90 is manufactured in a dedicated system. Typical pH: 9.5.