missing translation for 'onlineSavingsMsg'
Learn More

GOJO™ MICRELL™ 800 Series Bag-in-Box Dispenser

Catalog No. 19057607
Change view
Click to view available options
Volume (Metric):
800 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19057607 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19057607 Supplier GOJO™ Supplier No. 9721 12

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Push-style dispenser for MICRELL™ Antibacterial Lotion Soap

  • Compact size optimizes wall space
  • ADA compliant for away-from-sink placement
  • Lifetime guarantee
  • Compatible with MICRELL™ Bag-in-Box 800 mL refills

Specifications

Product Type Dispenser
Color White
For Use With (Application) Dispenser
For Use With (Equipment) MICRELL™ Antibacterial Lotion Soap
Quantity 12 Pk.
Volume (Metric) 800 mL