missing translation for 'onlineSavingsMsg'
Learn More

Michel's Fixative, Ricca Chemical

Catalog No. R5111000100
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R5111000100 100 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R5111000100 Supplier Ricca Chemical Company Supplier No. R5111000100A
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Michel's Fixative
Physical Form Liquid
CAS 7732-18-5, 7783-20-2, 6100-05-6, 10034-99-8, 128-53-0
CAS Min % 63.0, 34.57, 0.51, 0.08, 0.04
CAS Max % 65.0, 35.98, 0.53, 0.08, 0.04
Odor odorless to slight Ammonia odor
Packaging Natural Poly Bottle
Color Yellow
pH 7 to 7.2
Quantity 100 mL
Grade Laboratory
Show More Show Less