missing translation for 'onlineSavingsMsg'
Learn More

Intertek™ MIC Test Kit, Petroleum Microbiology Media: Acid Producing Bacteria Media

Catalog No. 08629021A
Encompass
Change view
Click to view available options
Quantity:
12 Vials
288 Vials
Packaging Type:
Vial
Product Type:
MIC Test Kit
Catalog No. Quantity Packaging Type Product Type
08629021A 12 Vials Vial MIC Test Kit
08629009C 288 Vials Vial MIC Test Kit
08629009A 288 Vials Vial MIC Test Kit
08629009B 288 Vials Vial MIC Test Kit
08629021B 12 Vials Vial MIC Test Kit
08629021C 12 Vials Vial MIC Test Kit
6 options

Catalog No. 08629021A

Supplier: Intertek™ MK106A

Only null left

Microbiology growth media products

A range of bacterial growth media and accessories for use in detecting and quantifying microbiologically influenced corrosion (MIC). Media products are available as boxes of 12 vials, cases of 288 vials, and in the single tube quick-test format.

Acid Producing Bacteria (APB) contribute to MIC by decreasing the pH of the environment which promotes the dissolution of metal.

  • APB: for quantification of acid producing bacteria and general heterotrophic anaerobic bacteria
  • The change in pH (production of acid) in APB-Q media is indicated by a change in color from red to yellow

Compliance

Manufactured in accordance with NACE Standard TM0194-94, Field Monitoring of Bacterial Growth in Oilfield Systems.

Specifications

Certifications/Compliance NACE, TM0194-94
Color Red
Manufacturing Origin United States
Product Type MIC Test Kit
For Use With (Application) Cultivation of Acid producing bacteria, MIC testing
Packaging Type Vial
Quantity 12 Vials