missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mexenone, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Pharmacopeia & Metrological Institutes Standards |
| CAS | 1641-17-4 |
| Flash Point | Not Applicable |
| Molecular Formula | C15H14O3 |
| Linear Formula | C15H14O3 |
| Quantity | 100 mg |
| Molecular Weight (g/mol) | 242.27 |
| Formula Weight | 242.27 |
| Grade | BP |
| Physical Form | Solid |