accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
Sign In
Don't have an account?
Create an Account
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
pH and Electrochemistry
pH and Electrochemistry Meter Accessories
Benchtop Titrator Accessories
361013093
Print
Mettler Toledo™ Replacement Parts for DV1000 Burets
Catalog No.
361013093
Change view
Chip view icon
List view icon
Click to view available options
For Use With (Equipment):
Mettler Toledo DV1000 Burets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
For Use With (Equipment)
361013093
Mettler Toledo DV1000 Burets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No.
361013093
Supplier
Mettler Toledo™
Supplier No.
23915
Sign In
or
Register
to check your price and availability.
$236.11
/
Each
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Specifications
Specifications
Type
Holder
For Use With (Equipment)
Mettler Toledo DV1000 Burets
Product Suggestions
Documents