missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Redox ORP Electrodes

An ORP/Redox electrode featuring a metal ring indicator designed for measuring oxidation/reduction potential; ideal for water and other general applications.

Supplier:  METTLER TOLEDO™ 51343203

 View more versions of this product

Catalog No. 01911503


Only null left
Explore available promotions

Description

Description

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Measuring oxidation/reduction potential (ORP)
  • Drinking water
  • Cooling towers
  • Swimming pools
  • Potable water supplies
  • Waste water
  • General lab

Accessories & Care

  • Ceramic junction (excluding the InLab Redox Flow with movable glass sleeve junction)
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • InLab Redox Pt805 and InLab Redox Ag805: no electrolyte required and must be paired with InLab Reference (51343190)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications

Specifications

S7
Glass
Combination Electrode
3mol/L KCl, AgCl saturated
S7 to BNC (30281915)
InLab Redox Micro
32°F to 212°F
3.93 in.
0.23 in.
Ceramic
0°C to 100°C
Platinum Ring Indicator
For measuring the oxidation/reduction power of solution
ARGENTHAL
3mol/L KCl
0°C to 100°C
100 mm
6 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions