missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ ISA Solutions

Catalog No. 01928356
Change view
Click to view available options
Type:
ISA Solution
ISS Solution
TISAB II with CDTA
TISAB III with CDTA
For Use With (Equipment):
Calcium Electrode
Fluoride Electrode
ISE Combination Electrode
Nitrate Electrode
Potassium Electrode
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
01928356 TISAB III with CDTA Fluoride Electrode
01928350 ISA Solution ISE Combination Electrode
01928351 ISA Solution Calcium Electrode
01928353 ISA Solution Nitrate Electrode
01928352 ISA Solution Potassium Electrode
01928354 ISS Solution Nitrate Electrode
01928355 TISAB II with CDTA Fluoride Electrode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01928356 Supplier Mettler Toledo™ Supplier No. 51344766

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Solutions for perfectION™ combined ion selective electrodes

Solutions (ionic-strength adjustment buffers)

  • Ion Electrolyte B (for chloride, copper, iodide, silver/sulfide ISE's), 475mL
  • ISA for calcium electrode, 475mL
  • ISA for potassium electrode, 475mL
  • ISA for nitrate electrode, 475mL
  • Nitrate ISS (for suppressing interference), 475mL
  • Fluoride TISAB II with CDTA, 3790mL
  • Fluoride TISAB III with CDTA (concentrate), 475mL

Specifications

Description perfectION TISAB III with CDTA
For Use With (Equipment) Fluoride Electrode
Type TISAB III with CDTA
Filling Solution Type Fluoride TISAB III with CDTA
Packaging 475 mL
Quantity 475 mL