missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Dairy pH Electrode

A pH electrode featuring a triple ceramic junction, designed for protein containing samples such as dairy products, milk and cream.

Supplier:  METTLER TOLEDO™ 59904591

Catalog No. 01911497


Only null left

Description

Description

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Milk and cream
  • Protein containing samples
  • Emulsion

Accessories & Care

  • Triple ceramic junction
  • A41 membrane glass
  • Refillable FRISCOLYT-B™ (51343185): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications

Specifications

pH electrode InLab Dairy
Triple/Ceramic
0 to 12
Combination Electrode
Used to puncture soft media or in aqueous samples
ARGENTHAL
InLab™ Dairy
0°F to 212°F
4.7 in.
0.47 in.
S7
Glass
0°C to 100°C
FRISCOLYT-B
S7 to BNC (30281915)
A41 Glass
3mol/L KCl
0°C to 100°C
120 mm
12 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions