Learn More
Mettler Toledo™ FillPal™ Sampling Pump
Catalog No. 10376206
Change view
Click to view available options
For Use With (Application):
Aggressive Samples (Acids, bases, solvents)
Catalog No. For Use With (Application)
10376206 Aggressive Samples (Acids, bases, solvents)
1 options
Catalog No. 10376206 Supplier Mettler Toledo™ Supplier No. 51337028
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Simplify the measurement of low viscosity samples. Mettler Toledo FillPal Sampling Pumps can be used for all DM density meters and RM refractometers.
  • Max. sample viscosity 1000mPa
  • Available in larger quantities 20mL or more
  • Suitable for all applications where the samples to be measured are of a similar nature and no intermediate complete drying of the measuring cell is required
  • Handles virtually all types of chemicals, including organic solvents, such as methyl ethyl ketone and toluene
Specifications
Product Type Sampling Pump
For Use With (Equipment) DM Density Meter and RM Refractometer
For Use With (Application) Aggressive Samples (Acids, bases, solvents)