missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Electrode Fill Solutions

Catalog No. 01911513
Encompass
Change view
Click to view available options
Filling Solution Type:
KCl saturated with AgCl; 25mL
For Use With (Equipment):
Reference Electrode
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filling Solution Type For Use With (Equipment)
01911513 KCl saturated with AgCl; 25mL Reference Electrode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01911513 Supplier Mettler Toledo™ Supplier No. 51343184
Only null left

Made from the highest quality reagents to provide optimum performance.

  • Aqueous solution containing inorganic and organic components
  • Supplied in 25mL (0.9 oz.) bottle

Specifications

Capacity (English) 0.9 oz.
Capacity (Metric) 25 mL
Concentration 3 mol/L
For Use With (Equipment) Reference Electrode
Filling Solution Type KCl saturated with AgCl; 25mL