missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ EasyPlus™ Titration Sensors

Catalog No. 01912527
Change view
Click to view available options
Type:
EG11-BNC Sensor pH aqueous
EG13-BNC sensor pH non-aqueous
Reference Electrolyte:
3mol/L KCl
LiCl, 1mol/L in ethanol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Reference Electrolyte
01912527 EG13-BNC sensor pH non-aqueous LiCl, 1mol/L in ethanol
01912526 EG11-BNC Sensor pH aqueous 3mol/L KCl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01912527 Supplier Mettler Toledo™ Supplier No. 30043104

Specially developed for use with the EasyPlus Titrators.

  • Used for routine acid/base application in aqueous media and non-aqueous solutions
  • Delivers quality and reliable results
  • Made of glass

pH aqueous Sensor

  • Ideal for acid/base titrations in aqueous media
  • Determines acidity in juice, milk and wine
  • Simultaneously measures m- and p-value in water
pH non-aqueous Sensor
  • Used in acid/base titrations in non-aqueous media such as free fatty acid content of edible oils

Specifications

Connector Types BNC Fixed Cable
Type EG13-BNC sensor pH non-aqueous
Internal Reference Conductor
Junction Ceramic
Material Glass
Reference Electrolyte LiCl, 1mol/L in ethanol
Temperature (English) Range 0°F to 140°F
Temperature (Metric) Range 0°C to 60°C
For Use With (Equipment) EasyPlus™ titrator