Learn More
Mettler Toledo™ EasyPlus™ Chloride, Redox mV, and Redox Ipol and Karl Fischer™ Sensors
Catalog No. 01912530
Change view
Click to view available options
For Use With (Equipment):
Chloride titration
Karl Fischer Redox titration
Redox titration
Temperature (English) Range:
0°F to 140°F
0°F to 212°F
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Temperature (English) Range
01912530 Chloride titration 0°F to 140°F
01912528 Karl Fischer Redox titration 0°F to 212°F
01912529 Redox titration 0°F to 140°F
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01912530 Supplier Mettler Toledo™ Supplier No. 30043107
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specially developed for use with the EasyPlus Titrators
  • Used for chloride precipitation, redox titrations/voltammetric/amperometric titrations
  • Delivers quality and reliable results
  • Made of glass
Chloride Sensor
  • Silver ring sensor for chloride titrations (especially for food and water)
  • Also used in silver content titrations
Redox mV Sensor
  • Platinum ring sensor for redox titrations (especially for food, electoplating or electronic industries)
  • Also used in applications like peroxide value or reducing sugar content
Redox Ipol and Karl Fischer Sensor
  • Simple, durable and zero-maintenance sensor primarily for Karl Fischer and redox titrations
  • For applications like important food ingredients such as Vitamin C or SO2 in wine
Specifications
Connector Types BNC Fixed Cable
Type Titration Sensor
Material Glass
Reference Electrolyte 1mol/L KCl
Temperature (English) Range 0°F to 140°F
Temperature (Metric) Range 0°C to 60°C
For Use With (Equipment) Chloride titration