Learn More
Mettler Toledo™ EasyPlus™ Chloride, Redox mV, and Redox Ipol and Karl Fischer™ Sensors
Specially developed for use with the EasyPlus Titrators
Supplier:  Mettler Toledo™ 30043106
 View more versions of this product
Catalog No. 01912529
Only null left
Description
Description
  • Used for chloride precipitation, redox titrations/voltammetric/amperometric titrations
  • Delivers quality and reliable results
  • Made of glass
Chloride Sensor
  • Silver ring sensor for chloride titrations (especially for food and water)
  • Also used in silver content titrations
Redox mV Sensor
  • Platinum ring sensor for redox titrations (especially for food, electoplating or electronic industries)
  • Also used in applications like peroxide value or reducing sugar content
Redox Ipol and Karl Fischer Sensor
  • Simple, durable and zero-maintenance sensor primarily for Karl Fischer and redox titrations
  • For applications like important food ingredients such as Vitamin C or SO2 in wine
Specifications
Specifications
BNC Fixed Cable
Glass
0°F to 140°F
Redox titration
Titration Sensor
3mol/L KCl
0°C to 60°C
Product Suggestions
Product Suggestions