missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Easy Na Sodium Analyzer: DX222-Na Sodium Half-Cell Electrode

Catalog No. 01912573
Change view
Click to view available options
Concentration Measurement Range:
1.0 to 1.0E-07 mol/L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration Measurement Range
01912573 1.0 to 1.0E-07 mol/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01912573 Supplier Mettler Toledo™ Supplier No. 30079616

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with Easy Na Sodium Analyzer.

  • For determining the sodium concentration of samples such as food with the Sodium Analyzer
  • Glass membrane with higher selectivity for sodium
  • Fast response time
  • No maintenance
  • Glass shaft
  • Measurement Range: 1.0 to 1.0E-07 mol/L
  • Optimal pH Range: 8 to 11
  • Temperature Range: 0° to 60°C

Specifications

Junction Glass Membrane
Temperature (Metric) Operating 0°C to 60°C
For Use With (Application) Determining the sodium concentration of samples such as food with the Sodium Analyzer.
Material Glass Membrane
pH Range 8 to 11
Concentration Measurement Range 1.0 to 1.0E-07 mol/L