missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Replacement Tubing for DL31 Karl Fischer™ Titrators

Replacement parts for use with DL31 Karl Fischer titrators.

$322.13

Specifications

Type Solvent Delivery/Drain Tube with Screw Fitting
Includes Solvent Dispensing Tubes, Suction Pipe
For Use With (Equipment) Mettler Toledo V20/V30 KF Titrators
View More Specs

Products 1
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
01911128
View Documents Promotion Details
Mettler Toledo™
51107481
Solvent Delivery/Drain Tube with Screw Fitting
Each for $322.13
Only null left
 
Description

Description

For DL31 Karl Fischer titrators.
Specifications

Specifications

Solvent Delivery/Drain Tube with Screw Fitting
Solvent Dispensing Tubes, Suction Pipe
Mettler Toledo V20/V30 KF Titrators
Videos
SDS
Documents

Documents