missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Redox Buffer Solutions
Storage and refill solutions for redox electrodes.
Supplier: Mettler Toledo™ 51350064
Description
Includes a quality inspection certificate which guarantees the stated values and traceability.Certifications
NIST traceable
Chemical Identifiers
| Buffer Solution | |
| Quality inspection certificate | |
| Completely Miscible |
Specifications
| Yellow | |
| Redox Buffer; UH/pH: 0.1 | |
| 468 mV | |
| 0.1 |
| Buffer Solution | |
| Quality inspection certificate | |
| 6 x 30 mL Bottle | |
| Completely Miscible |