Learn More
Metrohm 870 Titrino Plus Titrator
Supplier:  Metrohm KITTI870K20PS
Catalog No. 02-085-1039
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Titrator for volumetric water determination
  • Straightforward installation
  • Easy operation due to predefined methods
  • Excellent precision thanks to the high-resolution measuring input
  • Live curve for recognizing side-reactions
  • Overflow protection for increased operating safety
  • Compact USB printer as an option
Specifications
Specifications
RS232/USB
Volumetric water determination
20 mL
12.2 in.
5.5 in.
12.2 in.
Volumetric Titrator
Titration vessel and electrode
12.2 x 5.5 in. (310 x 142mm)
310 mm
142 mm
310 mm
Product Suggestions
Product Suggestions