Learn More
Metro™ Wire Shelving Track Sets
For use with Metro Super Erecta™, Super Erecta Pro™ and Super Adjustable™ Top Track™ shelving.
Supplier:  Metro™ TTS17NA
 View more versions of this product
Catalog No. 19-138-240
Only null left
Explore available promotions
Description
Description
  • Track sets include necessary sections of track for assembling track runs up to 21 ft. (640 cm)
  • Coated with Microban™ antimicrobial protection
Order Info
19-138-228 (TTS2.5NA) includes a joining kit to connect the track to other track sets.
Specifications
Specifications
Necessary sections of track for assembling track runs of 1830 to 6405mm
Track Set
518.2 cm
204 in.
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ top track shelving
Product Suggestions
Product Suggestions