Learn More
Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Wire shelves in stainless steel finish allow adjustable configurations when used with Metro Super Erecta shelving systems.
Supplier:  Metro™ 2454NS
 View more versions of this product
Catalog No. 19-030-983
Only null left
Explore available promotions
Description
Description
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Type 304 polished stainless steel
  • Used for the most aggressive applications and environments
  • Stands up to high temperature automated cart wash and autoclave applications
Specifications
Specifications
Wire Shelf
24 in.
61 x 137 in. (24 x 54 cm)
Plastic sleeves and wedges
Stainless Steel
610 mm
Metro Super Erecta Shelving System
Product Suggestions
Product Suggestions