Learn More
Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Wire shelves in stainless steel finish allow adjustable configurations when used with Metro Super Erecta shelving systems.
Supplier: Metro™ 2424NS
Description
- Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
- Type 304 polished stainless steel
- Used for the most aggressive applications and environments
- Stands up to high temperature automated cart wash and autoclave applications
Specifications
| Wire Shelf | |
| 24 in. | |
| 61 x 61 in. (24 x 24 cm) | |
| Plastic sleeves and wedges |
| Stainless Steel | |
| 610 mm | |
| Metro Super Erecta Shelving System |