Learn More
Metro™ Super Erecta™ Wire Shelf, Metroseal 3 with Microban™ Finish
Wire shelves are easily adjustable and the Metroseal 3 green epoxy with Microban™ finish promotes easy cleaning. For use with Metro Super Erecta shelving systems.
Supplier:  Metro™ 2442NK3
 View more versions of this product
Catalog No. 19-031-008
Only null left
Explore available promotions
Description
Description
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Corrosion resistant finish for wet or high humidity environments
  • Antimicrobial is built into finish to keep product clean
Specifications
Specifications
24 x 42 in.
Wire Shelf
Metroseal 3 with Microban
Product Suggestions
Product Suggestions