Learn More
Metro™ Super Erecta™ Wire Shelf, Metroseal 3 with Microban™ Finish
Wire shelves are easily adjustable and the Metroseal 3 green epoxy with Microban™ finish promotes easy cleaning. For use with Metro Super Erecta shelving systems.
Supplier: Metro™ 1830NK3
Description
- Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
- Corrosion resistant finish for wet or high humidity environments
- Antimicrobial is built into finish to keep product clean
Specifications
| 18 x 30 in. | |
| Wire Shelf | |
| Metroseal 3 with Microban |